Benzhydrol (Diphenylmethanol), LoGiCal, LGC Standards
Catalog # 77692-396
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:91-01-0
- Description:Benzhydrol (Diphenylmethanol)
- Size:10 mg
- Formula:C₁₃H₁₂O
- Molecular Weight:184.24 g/mol
- Cat. No.:77692-396