$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Clozapine, LoGiCal, LGC Standards
Clozapine, LoGiCal, LGC Standards
77692-394
:
LGC Standards
New Product
:
Clozapine, LoGiCal, LGC Standards
77692-394
:
LGC Standards
:
LGCFOR0480.00
New Product
:
CAS No.:
5786-21-0
Description:
Clozapine
Size:
10 mg
Formula:
C₁₈H₁₉ClN₄
Molecular Weight:
326.83 g/mol
Cat. No.:
77692-394