Order Entry
Puerto Rico
ContactUsLinkComponent
Trazodone Hydrochloride, LoGiCal, LGC Standards
Trazodone Hydrochloride, LoGiCal, LGC Standards
Supplier:  LGC Standards
New Product
Certificates
undefined
Trazodone Hydrochloride, LoGiCal, LGC Standards
Supplier:  LGC Standards
New Product

Specifications

  • CAS No.:
    25332-39-2
  • Formula:
    C₁₉H₂₃Cl₂N₅O
  • Molecular Weight:
    408.327 g/mol

Specifications

Frequently Bought Together