Order Entry
Puerto Rico
ContactUsLinkComponent
 
Chlortalidone, LoGiCal, LGC Standards
New Product
 
undefined
Chlortalidone, LoGiCal, LGC Standards
New Product

 

  • CAS No.:
    77-36-1
  • Formula:
    C₁₄H₁₁ClN₂O₄S
  • Molecular Weight:
    338.77 g/mol