Order Entry
Puerto Rico
ContactUsLinkComponent
Desmethylsildenafil (1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine), LoGiCal, LGC Standards
Desmethylsildenafil (1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine), LoGiCal, LGC Standards
Catalog # 77692-386
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Desmethylsildenafil (1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine), LoGiCal, LGC Standards
Catalog # 77692-386
Supplier:  LGC Standards
Supplier Number:  LGCFOR0254.08
New Product
CAS Number:  

Specifications

  • CAS No.:
    139755-82-1
  • Description:
    Desmethylsildenafil (1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine)
  • Size:
    10 mg
  • Formula:
    C₂₁H₂₈N₆O₄S
  • Molecular Weight:
    460.54 g/mol
  • Cat. No.:
    77692-386

Specifications

About this item