$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Chlorphenamine Maleate, LoGiCal, LGC Standards
Chlorphenamine Maleate, LoGiCal, LGC Standards
77692-374
:
LGC Standards
New Product
:
Chlorphenamine Maleate, LoGiCal, LGC Standards
77692-374
:
LGC Standards
:
LGCFOR0090.00
New Product
:
CAS No.:
113-92-8
Description:
Chlorphenamine Maleate
Size:
10 mg
Formula:
C₁₆H₁₉ClN₂
Molecular Weight:
390.86 g/mol
Cat. No.:
77692-374