$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Aciclovir, LoGiCal, LGC Standards
Aciclovir, LoGiCal, LGC Standards
77692-368
:
LGC Standards
New Product
:
Aciclovir, LoGiCal, LGC Standards
77692-368
:
LGC Standards
:
LGCFOR0061.00
New Product
:
CAS No.:
59277-89-3
Description:
Aciclovir
Size:
10 mg
Formula:
C₈H₁₁N₅O₃
Molecular Weight:
225.21 g/mol
Cat. No.:
77692-368