Dextromethorphan Hydrobromide Monohydrate, LoGiCal, LGC Standards
77692-367
New Product
:
- CAS No.:6700-34-1
- Description:Dextromethorphan Hydrobromide Monohydrate
- Size:10 mg
- Formula:C₁₈H₂₅NO·HBr·H₂O
- Molecular Weight:370.3 g/mol
- Cat. No.:77692-367