2-Amino-5-chlorobenzophenone, LoGiCal, LGC Standards
Catalog # 77692-366
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:719-59-5
- Description:2-Amino-5-chlorobenzophenone
- Size:10 mg
- Formula:C₁₃H₁₀ClNO
- Molecular Weight:231.68 g/mol
- Cat. No.:77692-366