Verapamil Hydrochloride, LoGiCal, LGC Standards
Catalog # 77693-679
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:152-11-4
- Description:Verapamil Hydrochloride
- Size:10 mg
- Formula:C₂₇H₃₈N₂O₄
- Molecular Weight:491.07 g/mol
- Cat. No.:77693-679