Order Entry
Puerto Rico
ContactUsLinkComponent
 
Cimetidine, LoGiCal, LGC Standards
New Product
 
undefined
Cimetidine, LoGiCal, LGC Standards
New Product

 

  • CAS No.:
    51481-61-9
  • Formula:
    C₁₀H₁₆N₆S
  • Molecular Weight:
    252.34 g/mol