(+)-Norpseudoephedrine Hydrochloride (Cathine Hydrochloride) 0.1 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-361
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2153-98-2
- Description:(+)-Norpseudoephedrine Hydrochloride (Cathine Hydrochloride) 0.1 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₉H₁₄ClNO
- Cat. No.:77692-361