Beclamide 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77692-360
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:501-68-8
- Description:Beclamide 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₁₀H₁₂ClNO
- Molecular Weight:197.661 g/mol
- Cat. No.:77692-360