Triflupromazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77693-677
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1098-60-8
- Description:Triflupromazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₈H₂₀ClF₃N₂S
- Molecular Weight:388.877 g/mol
- Cat. No.:77693-677