Order Entry
Puerto Rico
ContactUsLinkComponent
5-F-AKB-48 (5-F-APINACA; N-(1-Adamantyl)-1-(5-fluoropentyl)-1H-indazole-3-carboxamide) 0.1 mg/ml in Acetonitrile, LoGiCal, LGC Standards
5-F-AKB-48 (5-F-APINACA; N-(1-Adamantyl)-1-(5-fluoropentyl)-1H-indazole-3-carboxamide) 0.1 mg/ml in Acetonitrile, LoGiCal, LGC Standards
  77692-359
New Product
 :  
undefined
5-F-AKB-48 (5-F-APINACA; N-(1-Adamantyl)-1-(5-fluoropentyl)-1H-indazole-3-carboxamide) 0.1 mg/ml in Acetonitrile, LoGiCal, LGC Standards
  77692-359
 :  LGCAMP1396.13-12
New Product
 :  

 

  • CAS No.:
    1400742-13-3
  • Description:
    5-F-AKB-48 (5-F-APINACA; N-(1-Adamantyl)-1-(5-fluoropentyl)-1H-indazole-3-carboxamide) 0.1 mg/ml in Acetonitrile
  • Size:
    1 ml
  • Cat. No.:
    77692-359