Order Entry
Puerto Rico
ContactUsLinkComponent
UR-144 ((1-Pentylindol-3-yl)(2,2,3,3-tetramethyl-cyclopropyl)methanone; KM X-1) 0.1 mg/ml in Acetonitrile, LoGiCal, LGC Standards
UR-144 ((1-Pentylindol-3-yl)(2,2,3,3-tetramethyl-cyclopropyl)methanone; KM X-1) 0.1 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77693-676
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
UR-144 ((1-Pentylindol-3-yl)(2,2,3,3-tetramethyl-cyclopropyl)methanone; KM X-1) 0.1 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77693-676
Supplier:  LGC Standards
Supplier Number:  LGCAMP1396.10-12
New Product
CAS Number:  

Specifications

  • CAS No.:
    1199943-44-6
  • Description:
    UR-144 ((1-Pentylindol-3-yl)(2,2,3,3-tetramethyl-cyclopropyl)methanone; KM X-1) 0.1 mg/ml in Acetonitrile
  • Size:
    1 ml
  • Cat. No.:
    77693-676

Specifications

About this item