(S)-(-)-Methcathinone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77692-348
New Product
:
- CAS No.:66514-93-0
- Description:(S)-(-)-Methcathinone Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₀H₁₄ClNO
- Cat. No.:77692-348