Amobarbital 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-346
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:57-43-2
- Description:Amobarbital 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₁H₁₈N₂O₃
- Molecular Weight:226.28 g/mol
- Cat. No.:77692-346