Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Benzoylecgonine-D8 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Benzoylecgonine-D8 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Benzoylecgonine-D8 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77692-343
:
LGC Standards
New Product
:
Benzoylecgonine-D8 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77692-343
:
LGC Standards
:
LGCAMP1345.82-01
New Product
:
CAS No.:
205446-21-5
Description:
Benzoylecgonine-D8 1.0 mg/ml in Methanol
Size:
1 ml
Formula:
C₁₆H₁₁D₈NO₄
Cat. No.:
77692-343