Benzoylecgonine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-336
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:519-09-5
- Description:Benzoylecgonine 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₆H₁₉NO₄
- Cat. No.:77692-336