Clenbuterol Hydrochloride 1.0 mg/ml in Dimethyl Sulfoxide (as free base), LoGiCal, LGC Standards
77692-333
New Product
:
- CAS No.:21898-19-1
- Description:Clenbuterol Hydrochloride 1.0 mg/ml in Dimethyl Sulfoxide (as free base)
- Size:1 ml
- Formula:C₁₂H₁₈Cl₂N₂O
- Molecular Weight:313.65 g/mol
- Cat. No.:77692-333