1-Methyl-4-(phenylmethyl)piperazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-332
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:374898-00-7
- Description:1-Methyl-4-(phenylmethyl)piperazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₂H₁₉ClN₂
- Cat. No.:77692-332