2-Phenylethylamine Hydrochloride (Phenethylamine Hydrochloride; PEA HCl) 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-324
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:156-28-5
- Description:2-Phenylethylamine Hydrochloride (Phenethylamine Hydrochloride; PEA HCl) 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₈H₁₁N
- Molecular Weight:157.64 g/mol
- Cat. No.:77692-324