Clonazepam 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-307
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1622-61-3
- Description:Clonazepam 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₅H₁₀ClN₃O₃
- Cat. No.:77692-307