Allobarbital 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-303
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52-43-7
- Description:Allobarbital 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₀H₁₂N₂O₃
- Molecular Weight:208.22 g/mol
- Cat. No.:77692-303