Testosterone Propionate 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77693-669
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:57-85-2
- Description:Testosterone Propionate 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₂₂H₃₂O₃
- Molecular Weight:344.50 g/mol
- Cat. No.:77693-669