Amiodarone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-297
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:19774-82-4
- Description:Amiodarone Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₅H₂₉I₂NO₃
- Molecular Weight:681.78 g/mol
- Cat. No.:77692-297