Bupivacaine Hydrochloride Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal, LGC Standards
Catalog # 77692-294
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:73360-54-0
- Description:Bupivacaine Hydrochloride Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base)
- Size:1 ml
- Formula:C₁₈H₃₁ClN₂O₂
- Molecular Weight:342.908 g/mol
- Cat. No.:77692-294