Apomorphine Hydrochloride Hemihydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal, LGC Standards
Catalog # 77692-292
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:41372-20-7
- Description:Apomorphine Hydrochloride Hemihydrate 1.0 mg/ml in Methanol (as anhydrous free base)
- Size:1 ml
- Formula:C₁₇H₁₇NO₂·HCl·1/2H₂O
- Molecular Weight:312.79 g/mol
- Cat. No.:77692-292