Amiloride Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-291
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:17440-83-4
- Description:Amiloride Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₆H₈ClN₇O·HCl·2H₂O
- Molecular Weight:302.116 g/mol
- Cat. No.:77692-291