Clomipramine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-288
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:17321-77-6
- Description:Clomipramine Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₉H₂₄Cl₂N₂
- Molecular Weight:351.315 g/mol
- Cat. No.:77692-288