9-Hydroxyrisperidone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-287
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:144598-75-4
- Description:9-Hydroxyrisperidone 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₂₃H₂₇FN₄O₃
- Molecular Weight:426.49 g/mol
- Cat. No.:77692-287