Desmethylclozapine (Norclozapine) 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77692-286
New Product
:
- CAS No.:6104-71-8
- Description:Desmethylclozapine (Norclozapine) 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₇H₁₇ClN₄
- Molecular Weight:312.8 g/mol
- Cat. No.:77692-286