Trifluoperazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77693-661
New Product
:
- CAS No.:440-17-5
- Description:Trifluoperazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₁H₂₄F₃N₃S
- Molecular Weight:480.42 g/mol
- Cat. No.:77693-661