Timolol Maleate 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77693-660
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:26921-17-5
- Description:Timolol Maleate 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₃H₂₄N₄O₃S
- Molecular Weight:432.49 g/mol
- Cat. No.:77693-660