Acebutolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-284
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:34381-68-5
- Description:Acebutolol Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₈H₂₈N₂O₄·HCl
- Molecular Weight:372.89 g/mol
- Cat. No.:77692-284