Chlorpromazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-280
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:69-09-0
- Description:Chlorpromazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₇H₁₉ClN₂S
- Molecular Weight:355.32 g/mol
- Cat. No.:77692-280