Azelastine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77692-279
New Product
:
- CAS No.:79307-93-0
- Description:Azelastine Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₂H₂₄ClN₃O
- Molecular Weight:418.36 g/mol
- Cat. No.:77692-279