Testosterone 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77693-657
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:58-22-0
- Description:Testosterone 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₁₉H₂₈O₂
- Cat. No.:77693-657