Chlortalidone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-278
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:77-36-1
- Description:Chlortalidone 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₄H₁₁ClN₂O₄S
- Molecular Weight:338.77 g/mol
- Cat. No.:77692-278