Atropine 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77692-277
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:51-55-8
- Description:Atropine 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₁₇H₂₃NO₃
- Molecular Weight:289.38 g/mol
- Cat. No.:77692-277