$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Thebaine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Thebaine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77693-656
:
LGC Standards
New Product
:
Thebaine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77693-656
:
LGC Standards
:
LGCAMP0244.00-02
New Product
:
CAS No.:
115-37-7
Description:
Thebaine 0.1 mg/ml in Methanol
Size:
1 ml
Cat. No.:
77693-656