Order Entry
Puerto Rico
ContactUsLinkComponent
Digoxin 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Digoxin 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-274
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Digoxin 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-274
Supplier:  LGC Standards
Supplier Number:  LGCAMP0212.00-01
New Product
CAS Number:  

Specifications

  • CAS No.:
    20830-75-5
  • Description:
    Digoxin 1.0 mg/ml in Methanol
  • Size:
    1 ml
  • Formula:
    C₄₁H₆₄O₁₄
  • Molecular Weight:
    780.95 g/mol
  • Cat. No.:
    77692-274

Specifications

About this item