Antazoline Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77692-271
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2508-72-7
- Description:Antazoline Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₇H₁₉N₃
- Molecular Weight:301.82 g/mol
- Cat. No.:77692-271