alpha-Hydroxyalprazolam 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77692-266
New Product
:
- CAS No.:37115-43-8
- Description:alpha-Hydroxyalprazolam 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₇H₁₃CIN₄O
- Molecular Weight:324.8 g/mol
- Cat. No.:77692-266