Order Entry
Puerto Rico
ContactUsLinkComponent
Alprazolam 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Alprazolam 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-265
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Alprazolam 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-265
Supplier:  LGC Standards
Supplier Number:  LGCAMP0136.00-02
New Product
CAS Number:  

Specifications

  • CAS No.:
    28981-97-7
  • Description:
    Alprazolam 0.1 mg/ml in Methanol
  • Size:
    1 ml
  • Formula:
    C₁₇H₁₃ClN₄
  • Molecular Weight:
    308.76 g/mol
  • Cat. No.:
    77692-265

Specifications

About this item