$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Buprenorphine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Buprenorphine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77692-261
:
LGC Standards
New Product
:
Buprenorphine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77692-261
:
LGC Standards
:
LGCAMP0120.00-02
New Product
:
CAS No.:
52485-79-7
Description:
Buprenorphine 0.1 mg/ml in Methanol
Size:
1 ml
Cat. No.:
77692-261