Bromazepam 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-245
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1812-30-2
- Description:Bromazepam 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₄H₁₀BrN₃O
- Cat. No.:77692-245