Aciclovir 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal, LGC Standards
77692-244
New Product
:
- CAS No.:59277-89-3
- Description:Aciclovir 1.0 mg/ml in Dimethyl Sulfoxide
- Size:1 ml
- Formula:C₈H₁₁N₅O₃
- Molecular Weight:225.21 g/mol
- Cat. No.:77692-244