Dextromethorphan Hydrobromide Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal, LGC Standards
Catalog # 77692-241
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:6700-34-1
- Description:Dextromethorphan Hydrobromide Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base)
- Size:1 ml
- Formula:C₁₈H₂₅NO·HBr·H₂O
- Molecular Weight:370.3 g/mol
- Cat. No.:77692-241